missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, 99%, extra pure

Product Code. 10457370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10457370 2.5 L Glass Bottle 2.5 L
10447370 1 L Glass Bottle 1 L
10329093 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10457370 Supplier Thermo Scientific Acros Supplier No. 156020025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-80-5 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
IUPAC Name 2-ethoxyethanol
SMILES CCOCCO

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70°C
Density 0.9300g/mL
Boiling Point 135.0°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula C2H5OCH2CH2OH
Refractive Index 1.4060 to 1.4100
Quantity 2.5 L
Beilstein 01, 467
Merck Index 15, 3803
Specific Gravity 0.93
Solubility Information Solubility in water: complete solubility. Other solubilities: miscible with most organic solvents
Viscosity 2.08 mPa.s (20°C)
Formula Weight 90.12
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethanol
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • 2-Ethoxyethanol
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Indicaciones de peligro
  • H360FD-May damage fertility. May damage the unborn child.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.