missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, ≥99%, Spectrophotometric Grade, Honeywell™ Riedel-de-Haën™

Product Code. 15602770
Change view
Click to view available options
Quantity:
2 L
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15602770 2 L 2 L
15692760 2 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15602770 Supplier Honeywell Riedel-de Haën Supplier No. 2563742L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
IUPAC Name 2-ethoxyethan-1-ol
SMILES CCOCCO

Specifications

Density 0.93g/mL at 25°C
Explosion Limit 0.14
Flash Point 42°C (107.6°F)
Ignition Point 460°F
Linear Formula C2H5OCH2CH2OH
Quantity 2 L
UN Number UN1171
Beilstein 1098271
Vapor Density 3.1 (vs air)
Formula Weight 90.12g/mol
Vapor Pressure 3.8mmHg (20°C)
Percent Purity ≥99%
Grade Sprectrophotometric
Chemical Name or Material 2-Ethoxyethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.