missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, TRC

Product Code. 30005948
Change view
Click to view available options
Quantity:
5 mL
100 mL
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005948 100 mL 100mL
30138127 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005948 Supplier Toronto Research Chemicals Supplier No. TRCE678265100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-80-5
Formula Weight 90.0681
InChI Formula InChI=1S/C4H10O2/c1-2-6-4-3-5/h5H,2-4H2,1H3
IUPAC Name 2-ethoxyethanol
Molecular Formula C4 H10 O2
Molecular Weight (g/mol) 90.12
SMILES CCOCCO
Synonym 2-Ethoxyethanol, Ethanol, 2-ethoxy-, 2-Ethoxyethanol, 2-Ethoxyethan-1-ol, 2-Ethoxyethyl alcohol, Bikanol E 1, Cellosolve, Dowanol EE, Ektasolve EE, Emkanol, Ethoxyethanol, Ethyl 2-hydroxyethyl ether, Ethyl Cellosolve, Ethylene glycol ethyl ether, Ethylene glycol monoethyl ether, Glycol monoethyl ether, Monoethylene glycol ethyl ether, NSC 8837, Oxitol, Poly-Solv EE, Seahosol MG, Shihozoru MG, Solvid, β-Ethoxyethanol
Chemical Name or Material 2-Ethoxyethanol

Specifications

Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.