missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl Acetate, 98+%, Pure, Fisher Chemical

Product Code. 10111810 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10111810 2.5 L Plastic Coated Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10111810 Supplier Fisher Chemical Supplier No. E/0800/PB17
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 111-15-9
Melting Point -61°C
Color Yellow
Boiling Point 156°C
Packaging Plastic Coated Glass bottle
Molecular Formula C6H12O3
MDL Number MFCD00009251
Quantity 2.5 L
Synonym ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu
InChI Key SVONRAPFKPVNKG-UHFFFAOYSA-N
SMILES CCOCCOC(C)=O
IUPAC Name 2-ethoxyethyl acetate
Molecular Weight (g/mol) 132.16
PubChem CID 8095
Formula Weight 132.16g/mol
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.