missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyphenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10702133
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10702133 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10702133 Supplier Thermo Scientific Maybridge Supplier No. TL00819EA
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 23163-84-0
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C9H9NOS
Linear Formula CH3CH2OC6H4NCS
MDL Number MFCD00046793
Quantity 10 g
Synonym 2-ethoxyphenyl isothiocyanate, o-ethoxyphenyl isothiocyanate, 2-ethoxyphenylisothiocyanate, 1-ethoxy-2-isothiocyanato-benzene, benzene,1-ethoxy-2-isothiocyanato, benzene, 1-ethoxy-2-isothiocyanato, 2-ethoxybenzenisothiocyanate, 2-isothiocyanatophenetole, ethoxyphenyl isothiocyanate, 3-trimethylsilyl-2-propene-1-ol
InChI Key MAHMRCJOZYLGOW-UHFFFAOYSA-N
SMILES CCOC1=CC=CC=C1N=C=S
IUPAC Name 1-ethoxy-2-isothiocyanatobenzene
Molecular Weight (g/mol) 179.237
PubChem CID 123407
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 2-Ethoxyphenyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.