missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-butene, 97%

Product Code. 11372487
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372487 25 g 25g
11362487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372487 Supplier Thermo Scientific Alfa Aesar Supplier No. L11553.14
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 760-21-4 | C6H12 | 84.162 g/mol

2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in alcohol, and methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 760-21-4
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009341
InChI Key RYKZRKKEYSRDNF-UHFFFAOYSA-N
Synonym 2-ethyl-1-butene, 3-methylenepentane, pentane, 3-methylene, 2-ethylbut-1-ene, 1-butene, 2-ethyl, unii-o5mh5v756e, 1,1-diethylethene, acmc-1blm9, 1-butene, 2-ethyl-8ci
PubChem CID 12970
IUPAC Name 3-methylidenepentane
SMILES CCC(=C)CC

Specifications

Melting Point -132°C
Density 0.689
Boiling Point 63°C to 64°C
Flash Point −26°C (−14°F)
Refractive Index 1.396
Quantity 25 g
UN Number UN3295
Beilstein 1697102
Solubility Information Soluble in alcohol,and methanol. Insoluble in water.
Formula Weight 84.16
Percent Purity 97%
Chemical Name or Material 2-Ethyl-1-butene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.