missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-hexanol, 99%

Product Code. 10102580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10102580 1 L Glass Bottle 1 L
10773851 5 mL Glass Bottle 5mL
10488450 2.5 L Glass Bottle 2.5 L
10063803 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10102580 Supplier Thermo Scientific Acros Supplier No. 118530010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-76-7 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-76-7
MDL Number MFCD00004746
InChI Key YIWUKEYIRIRTPP-UHFFFAOYSA-N
Synonym 2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151
PubChem CID 7720
ChEBI CHEBI:16011
IUPAC Name 2-ethylhexan-1-ol
SMILES CCCCC(CC)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -76.0°C
Density 0.8330g/mL
Boiling Point 184.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H18O
Linear Formula CH3(CH2)3CH(C2H5)CH2OH
Refractive Index 1.4300 to 1.4330
Quantity 1 L
Merck Index 15, 3863
Specific Gravity 0.833
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in many organic solvents including, ethyl alcohol and ethyl ether
Molecular Weight (g/mol) 130.23
Viscosity 10 mPa.s (20°C)
Formula Weight 130.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-hexanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethyl-1-hexanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Ethylhexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.