missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-Methylsuccinic Acid, MP Biomedicals™

Product Code. 15717909 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15717909 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15717909 Supplier MP Biomedicals Supplier No. 05205867.6
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2-Ethyl-2-Methylsuccinic Acid

Specifications

CAS 631-31-2
Melting Point 100°C to 102°C
Color White
Molecular Formula C7H12O4
MDL Number MFCD00004201
Quantity 50 mg
Synonym Isopimelic Acid, α-Ethyl methyl succinic acid
InChI Key FDYJJKHDNNVUDR-UHFFFAOYNA-N
Molecular Weight (g/mol) 160.17
Percent Purity >98%
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.