missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-methylsuccinic Acid, TRC

Product Code. 30380340
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380340 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380340 Supplier Toronto Research Chemicals Supplier No. TRCE817970100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethyl-2-Methylsuccinic Acid
CAS 631-31-2
Molecular Formula C7 H12 O4
Quantity 100 mg
Synonym Butanedioic acid, 2-ethyl-2-methyl-, Succinic acid, 2-ethyl-2-methyl- (6CI,7CI,8CI), 2-Ethyl-2-methylsuccinic acid, 2-Methyl-2-ethylsuccinic acid, 3-Methyl-3-carboxyvaleric acid, NSC 38752, α-Methyl-α-ethylsuccinic acid, (2RS)-2-Ethyl-2-methylbutanedioic acid
InChI Formula InChI=1S/C7H12O4/c1-3-7(2,6(10)11)4-5(8)9/h3-4H2,1-2H3,(H,8,9)(H,10,11)
SMILES CCC(C)(CC(=O)O)C(=O)O
IUPAC Name 2-ethyl-2-methylbutanedioic acid
Molecular Weight (g/mol) 160.168
Formula Weight 160.074
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.