missing translation for 'onlineSavingsMsg'
Learn More

2'-Ethyl Simvastatin, TRC

Product Code. 30012784
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012784 10 mg 10mg
18073550 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012784 Supplier Toronto Research Chemicals Supplier No. TRCE92498510MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79902-42-4
Molecular Formula C23 H34 O5
Molecular Weight (g/mol) 390.51
Synonym Propanoic acid, 2-methyl-, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, Propanoic acid, 2-methyl-, 1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester, [1S-[1α,3α,7β,8β(2S*,4S*),8aβ]]- (9CI), 2'-Ethyl Simvastatin, [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-Hydroxy-6-oxo-tetrahydropyran-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2-methylpropanoate, (1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-Hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2-methylpropanoate, 2'-Desethyl Simvastatin, Simvastatin Impurity I (EP)
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2-methylpropanoate
SMILES CC(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H34O5/c1-13(2)23(26)28-20-10-14(3)9-16-6-5-15(4)19(22(16)20)8-7-18-11-17(24)12-21(25)27-18/h5-6,9,13-15,17-20,22,24H,7-8,10-12H2,1-4H3/t14-,15-,17+,18+,19-,20-,22-/m0/s1
Formula Weight 390.2406
Chemical Name or Material 2'-Desethyl Simvastatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.