missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylamino)ethanol, 98%

Product Code. 11412398
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412398 2500 mL 2500mL
11492388 100 mL 100mL
11402398 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412398 Supplier Thermo Scientific Alfa Aesar Supplier No. A15744.0F
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-73-6 | C4H11NO | 89.138 g/mol

2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.

Solubility
Miscible in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-73-6
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002841
InChI Key MIJDSYMOBYNHOT-UHFFFAOYSA-N
Synonym 2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
PubChem CID 8072
IUPAC Name 2-(ethylamino)ethanol
SMILES CCNCCO

Specifications

Melting Point -9°C
Density 0.914
Boiling Point 168°C to 170°C
Flash Point 71°C (160°F)
Linear Formula CH3CH2NHCH2CH2OH
Refractive Index 1.441
Quantity 2500 mL
UN Number UN2922
Beilstein 635671
Solubility Information Miscible in water.
Formula Weight 89.14
Percent Purity 98%
Chemical Name or Material 2-(Ethylamino)ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Ethylamino)ethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Ethylaminoethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.