missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylamino)ethanol, 98%

Product Code. 10625802 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10625802 1 kg Glass bottle 1kg
10002080 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10625802 Supplier Thermo Scientific Acros Supplier No. 168730010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-73-6 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-73-6
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00002841
InChI Key MIJDSYMOBYNHOT-UHFFFAOYSA-N
Synonym 2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
PubChem CID 8072
IUPAC Name 2-(ethylamino)ethanol
SMILES CCNCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -6°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 167°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5NHCH2CH2OH
Refractive Index 1.4390 to 1.441
Quantity 1 kg
Beilstein 04, 282
Specific Gravity 0.91
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol and ether
Formula Weight 89.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Ethylamino)ethanol, 98%
Show More Show Less

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.