missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylfuran, 98+%

Product Code. 11492927
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492927 100 g 100g
11472927 5 g 5g
11482927 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492927 Supplier Thermo Scientific Alfa Aesar Supplier No. A15520.22
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3208-16-0 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3208-16-0
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00003259
InChI Key HLPIHRDZBHXTFJ-UHFFFAOYSA-N
Synonym furan, 2-ethyl, alpha-ethylfuran, 2-ethyloxole, 2-ethyl furan, ccris 4387, unii-t8o6j71t9o, fema no. 3673, furan, .alpha.-ethyl, a-ethylfuran, 2-ethyl furane
PubChem CID 18554
IUPAC Name 2-ethylfuran
SMILES CCC1=CC=CO1

Specifications

Density 0.912
Boiling Point 92°C to 93°C
Flash Point −2°C (28°F)
Refractive Index 1.44
Quantity 100 g
UN Number UN3271
Beilstein 105401
Sensitivity Air Sensitive
Formula Weight 96.13
Percent Purity ≥98%
Chemical Name or Material 2-Ethylfuran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.