missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanoic acid, 99%

Product Code. 10276663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10276663 1 kg Glass Bottle 1kg
10559600 500 g Glass Bottle 500g
10653702 2.5 kg Plastic drum 2.5kg
11944931 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10276663 Supplier Thermo Scientific Acros Supplier No. 118520011
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-57-5 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-57-5
MDL Number MFCD00002675
InChI Key OBETXYAYXDNJHR-UHFFFAOYSA-N
Synonym 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid
PubChem CID 8697
IUPAC Name 2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -59.0°C
Density 0.9060g/mL
Boiling Point 228.0°C
Flash Point 114°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H16O2
Linear Formula CH3(CH2)3CH(C2H5)CO2H
Refractive Index 1.4240 to 1.4260
Quantity 1 kg
Beilstein 02,349
Specific Gravity 0.906
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in ether
Molecular Weight (g/mol) 144.21
Viscosity 7.7 mPa.s (20°C)
Formula Weight 144.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexanoic acid
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.