missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Acetate, 99%, Honeywell™

Product Code. 15615780 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15615780 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15615780 Supplier Honeywell Research Chemicals Supplier No. 5374971L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 103-09-3
Melting Point -92°C
Color Colorless
Density 0.87g/mL at 25°C
Boiling Point 199°C
Flash Point 71°C (159.8°F)
Packaging Glass bottle
Molecular Formula C10H20O2
Linear Formula H3CCO2CH2CH(C2H5)(CH2)3CH3
MDL Number MFCD00027249
Quantity 1 L
Synonym acetic acid, 2-ethylhexyl ester, 2-ethyl-1-hexanol acetate, 2-ethylhexyl ethanoate, 2-ethyl-1-hexyl acetate, 2-ethylhexanyl acetate, 2-ethylhexylacetate, beta-ethylhexyl acetate, acetic acid 2-ethylhexyl ester, fema number 2806, 2-ethylhexylester kyseliny octove
Solubility Information 0.0039g/L at 20°C Solubility in water
InChI Key WOYWLLHHWAMFCB-UHFFFAOYNA-N
SMILES CCCCC(CC)COC(C)=O
IUPAC Name 2-ethylhexyl acetate
Molecular Weight (g/mol) 172.27
PubChem CID 7635
ChEBI CHEBI:87392
Formula Weight 172.26g/mol
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.