missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl acetate, 99%, pure

Product Code. 10685861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10685861 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10685861 Supplier Thermo Scientific Acros Supplier No. 410105000
  • ONLINE EXCLUSIVE PRICE
    57.40 €
    55.70 € / 500g
    Save 1.70 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 103-09-3 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 103-09-3
CAS Min % 98.5
CAS Max % 100.0
Melting Point -45°C
Color Colorless
Density 0.8700g/mL
Boiling Point 197°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C10H20O2
Linear Formula CH3COOCH2CH(C2H5)(CH2)3CH3
Refractive Index 1.4190 to 1.421
MDL Number MFCD00027249
Quantity 500 g
Merck Index 15, 6852
Specific Gravity 0.87
Synonym acetic acid, 2-ethylhexyl ester, 2-ethyl-1-hexanol acetate, 2-ethylhexyl ethanoate, 2-ethyl-1-hexyl acetate, 2-ethylhexanyl acetate, 2-ethylhexylacetate, beta-ethylhexyl acetate, acetic acid 2-ethylhexyl ester, fema number 2806, 2-ethylhexylester kyseliny octove
Solubility Information Solubility in water: insoluble
InChI Key WOYWLLHHWAMFCB-UHFFFAOYNA-N
SMILES CCCCC(CC)COC(C)=O
IUPAC Name 2-ethylhexyl acetate
Molecular Weight (g/mol) 172.27
PubChem CID 7635
ChEBI CHEBI:87392
Formula Weight 172.27
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl acetate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethylhexyl acetate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.