missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl acetate, 99%

Product Code. 11470857
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470857 250 mL 250mL
11460857 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470857 Supplier Thermo Scientific Alfa Aesar Supplier No. B23257.AK
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-09-3 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-09-3
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00027249
InChI Key WOYWLLHHWAMFCB-UHFFFAOYNA-N
Synonym acetic acid, 2-ethylhexyl ester, 2-ethyl-1-hexanol acetate, 2-ethylhexyl ethanoate, 2-ethyl-1-hexyl acetate, 2-ethylhexanyl acetate, 2-ethylhexylacetate, beta-ethylhexyl acetate, acetic acid 2-ethylhexyl ester, fema number 2806, 2-ethylhexylester kyseliny octove
PubChem CID 7635
ChEBI CHEBI:87392
IUPAC Name 2-ethylhexyl acetate
SMILES CCCCC(CC)COC(C)=O

Specifications

Melting Point -45°C
Density 0.87
Boiling Point 199°C to 200°C
Flash Point 76°C (169°F)
Odor Pleasant
Refractive Index 1.42
Quantity 250 mL
Beilstein 1758321
Merck Index 14,6763
Formula Weight 172.27
Percent Purity 99%
Chemical Name or Material 2-Ethylhexyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.