missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl acrylate, 98%, stab.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylhexyl acrylate is used in the preparation of homopolymers and copolymers. It is involved in the copolymerization with acrylamide, methacrylic acid, acrylonitrile, styrene, maleic acid esters, vinyl acetate, vinyl chloride and unsaturated polyesters. Further, it acts as a starting material in chemical syntheses.
Solubility
Miscible with ether and alcohol. Immiscible with water.
Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
2-Ethylhexyl acrylate is used in the preparation of homopolymers and copolymers. It is involved in the copolymerization with acrylamide, methacrylic acid, acrylonitrile, styrene, maleic acid esters, vinyl acetate, vinyl chloride and unsaturated polyesters. Further, it acts as a starting material in chemical syntheses.
Solubility
Miscible with ether and alcohol. Immiscible with water.
Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | -90°C |
| Density | 0.885 |
| Boiling Point | 130°C (50 mmHg) |
| Flash Point | 79°C (174°F) |
| Refractive Index | 1.436 |
| Quantity | 100 mL |
| Beilstein | 1765828 |
| Solubility Information | Miscible with ether and alcohol. Immiscible with water. |
| Formula Weight | 184.28 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
Product Identifier
- 2-Ethylhexyl acrylate, stabilized
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contain: 2-Ethylhexyl acrylate, 4-Methoxyphenol
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?