missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl acrylate, 98%, stab.

Product Code. 11346676
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346676 500 mL 500mL
11336676 100 mL 100mL
11356676 2500 mL 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346676 Supplier Thermo Scientific Alfa Aesar Supplier No. L03854.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-11-7 | C11H20O2 | 184.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexyl acrylate is used in the preparation of homopolymers and copolymers. It is involved in the copolymerization with acrylamide, methacrylic acid, acrylonitrile, styrene, maleic acid esters, vinyl acetate, vinyl chloride and unsaturated polyesters. Further, it acts as a starting material in chemical syntheses.

Solubility
Miscible with ether and alcohol. Immiscible with water.

Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-11-7
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00009495
InChI Key GOXQRTZXKQZDDN-UHFFFAOYSA-N
Synonym 2-ethylhexyl acrylate, 2-propenoic acid, 2-ethylhexyl ester, 2-ethyl-1-hexyl acrylate, 2-ethylhexyl 2-propenoate, acrylic acid, 2-ethylhexyl ester, 2-ethylhexylacrylate, acrylic acid 2-ethylhexyl ester, 1-hexanol, 2-ethyl-, acrylate, ccris 3430, 2-ethylhexylester kyseliny akrylove
PubChem CID 7636
ChEBI CHEBI:82465
IUPAC Name 2-ethylhexyl prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=C

Specifications

Melting Point -90°C
Density 0.885
Boiling Point 130°C (50 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.436
Quantity 500 mL
Beilstein 1765828
Solubility Information Miscible with ether and alcohol. Immiscible with water.
Formula Weight 184.28
Percent Purity 98%
Chemical Name or Material 2-Ethylhexyl acrylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.