missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl acrylate, 99+%, stabilized

Product Code. 10771704 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
2.5kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10771704 25 g Glass bottle 25g
10341731 1 kg Glass bottle 1kg
10432975 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10771704 Supplier Thermo Scientific Acros Supplier No. 410110250
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-11-7 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-11-7 , 150-76-5
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00009495
InChI Key GOXQRTZXKQZDDN-UHFFFAOYSA-N
Synonym 2-ethylhexyl acrylate, 2-propenoic acid, 2-ethylhexyl ester, 2-ethyl-1-hexyl acrylate, 2-ethylhexyl 2-propenoate, acrylic acid, 2-ethylhexyl ester, 2-ethylhexylacrylate, acrylic acid 2-ethylhexyl ester, 1-hexanol, 2-ethyl-, acrylate, ccris 3430, 2-ethylhexylester kyseliny akrylove
PubChem CID 7636
ChEBI CHEBI:82465
IUPAC Name 2-ethylhexyl prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=C

Specifications

Acidity 0.01% max. (as acrylic acid)
CAS Min % 99.0
CAS Max % 100.0
Melting Point -90.0°C
Density 0.8800g/mL
Boiling Point 215.0°C to 219.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH2=CHCOOCH2CH(CH2CH3)(CH2)3CH3
Refractive Index 1.4340 to 1.4360
Quantity 25 g
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble
Stabilizer 10 to 20ppm MEHQ
Viscosity 1.7 mPa.s (20°C)
Formula Weight 184.28
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl acrylate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethylhexyl acrylate, stabilized
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.