missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl methacrylate, 99%, stabilized

Produktkod. 10242482 Handla allt Thermo Scientific Chemicals Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Förpackningsstorlek:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging unitSize
10242482 1 L Glass bottle 1 L
10671853 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10242482 Leverantör Thermo Scientific Acros Leverantörsnummer 371170010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 688-84-6 | C12H22O2 | 198.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 688-84-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.31
MDL Number MFCD00009494
InChI Key WDQMWEYDKDCEHT-UHFFFAOYSA-N
Synonym 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
PubChem CID 12713
IUPAC Name 2-ethylhexyl 2-methylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(=C)C

Specifikationer

CAS Min % 98.5
CAS Max % 100.0
Melting Point -50.0°C
Density 0.8840g/mL
Boiling Point 120°C (18.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CH(C2H5)(CH2)3CH3
Refractive Index 1.4370 to 1.4390
Quantity 1 L
Specific Gravity 0.884
Solubility Information Solubility in water: 0.0031g/L
Stabilizer 40 to 60ppm MEHQ
Formula Weight 198.31
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl methacrylate
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 2-Ethylhexyl methacrylate, stabilized
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.