missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Salicylate, TRC

Product Code. 30384722
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384722 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384722 Supplier Toronto Research Chemicals Supplier No. TRCE91877550G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 118-60-5
Molecular Formula C15 H22 O3
Quantity 50 g
Synonym 2-Hydroxybenzoic acid 2-ethylhexyl ester, 2-Ethylhexyl o-hydroxybenzoate, 2-Ethylhexyl salicylate, Dermoblock OS, Escalol 587, Eusolex OS, HallBrite OS-USP, NSC 46151, Neo Heliopan OS, Octisalate, Octyl salicylate, Parsol EHS, Salicylic acid, 2-ethylhexyl ester, Solarom OS, Sunarome O, Sunarome WMO, Uvinul 0-18, WMO
InChI Formula InChI=1S/C15H22O3/c1-3-5-8-12(4-2)11-18-15(17)13-9-6-7-10-14(13)16/h6-7,9-10,12,16H,3-5,8,11H2,1-2H3
SMILES CCCCC(CC)COC(=O)c1ccccc1O
IUPAC Name 2-ethylhexyl 2-hydroxybenzoate
Molecular Weight (g/mol) 250.33
Formula Weight 250.1569
Chemical Name or Material Octisalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.