Learn More
2-Ethylhexylamine, 98%
Description
2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.
Solubility
Soluble in water. 2.5 g/L (20°C)
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Protect against electrostatic charges. Store away from acid chlorides and acid anhydrides.
Specifications
Specifications
| Melting Point | -76°C |
| Density | 0.789 |
| Boiling Point | 169°C |
| Flash Point | 50°C (122°F) |
| Odor | Amine-like |
| Refractive Index | 1.43 |
| Quantity | 100 mL |
| UN Number | UN2276 |
| Beilstein | 1209249 |
| Sensitivity | Air Sensitive |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.