missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10432863
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432863 1 g Amber Glass Bottle 1g
10773137 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10432863 Supplier Thermo Scientific Maybridge Supplier No. AC31420DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40411-25-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00002013
InChI Key ZVFNUQWYLXXSJM-UHFFFAOYSA-N
Synonym 2-ethylphenyl isocyanate, 2-ethylphenylisocyanate, o-ethylphenyl isocyanate, benzene, ethylisocyanato, 2-ethylbenzenisocyanate, 2-eth-ylphenyl isocyanate, 2-ethyl phenyl isocyanate, 2-ethyl-phenyl isocyanate, acmc-1amc4, 1-ethyl-2-isocyanato-benzene
PubChem CID 583486
IUPAC Name 1-ethyl-2-isocyanatobenzene
SMILES CCC1=CC=CC=C1N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 1.04g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH2C6H4NCO
Quantity 1 g
Formula Weight 147.18
Chemical Name or Material 2-Ethylphenyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.