missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenyl isocyanate, 99%

Product Code. 15499019
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499019 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15499019 Supplier Thermo Scientific Alfa Aesar Supplier No. L15252.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40411-25-4 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 40411-25-4
Density 1.043
Boiling Point 56°C (3 mmHg)
Flash Point 78°C (172°F)
Molecular Formula C9H9NO
Refractive Index 1.528
MDL Number MFCD00002013
Quantity 5 g
UN Number UN2206
Beilstein 3538910
Sensitivity Moisture sensitive
Synonym 2-ethylphenyl isocyanate, 2-ethylphenylisocyanate, o-ethylphenyl isocyanate, benzene, ethylisocyanato, 2-ethylbenzenisocyanate, 2-eth-ylphenyl isocyanate, 2-ethyl phenyl isocyanate, 2-ethyl-phenyl isocyanate, acmc-1amc4, 1-ethyl-2-isocyanato-benzene
InChI Key ZVFNUQWYLXXSJM-UHFFFAOYSA-N
SMILES CCC1=CC=CC=C1N=C=O
IUPAC Name 1-ethyl-2-isocyanatobenzene
Molecular Weight (g/mol) 147.177
PubChem CID 583486
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 2-Ethylphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.