missing translation for 'onlineSavingsMsg'
Learn More

2-(ethylsulfanyl)ethane-1-thiol, TRC

Product Code. 30374497
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374497 50 mg
30387225 100 mg
30381049 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374497 Supplier Toronto Research Chemicals Supplier No. TRCE90232350MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26750-44-7
Molecular Formula C4H10S2
Molecular Weight (g/mol) 122.25
IUPAC Name 2-ethylsulfanylethanethiol
SMILES CCSCCS

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C4H10S2/c1-2-6-4-3-5/h5H,2-4H2,1H3
Formula Weight 122.0224
Chemical Name or Material 2-(ethylsulfanyl)ethane-1-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.