missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylthio-1,3,4-thiadiazole-5-thiol, 97%, Thermo Scientific Chemicals

Product Code. 11420637
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420637 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11420637 Supplier Thermo Scientific Alfa Aesar Supplier No. B22464.06
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37147-15-2
Molecular Formula C4H6N2S3
Molecular Weight (g/mol) 178.29
MDL Number MFCD00186131
InChI Key JLJYQXMUUKOBBO-UHFFFAOYSA-N
Synonym 2-ethylthio-1,3,4-thiadiazole-5-thiol, 5-ethylthio-1,3,4-thiadiazole-2-thiol, 5-ethylsulfanyl-1,3,4-thiadiazole-2-thiol, 2-ethylthio-1,3,4-thiazole-5-thiol, 5-ethylthio-1,3,4-thiadiazole-2 3h-thione, 5-ethylsulfanyl-1,3,4-thiadiazole-2 3h-thione, acmc-1cszq, 5-ethylthio-3h-1,3,4-thiadiazole-2-thione
PubChem CID 1201366
IUPAC Name 5-ethylsulfanyl-3H-1,3,4-thiadiazole-2-thione
SMILES CCSC1=NNC(=S)S1

Specifications

Melting Point 119°C to 122°C
Quantity 5 g
Formula Weight 178.3
Percent Purity 97%
Chemical Name or Material 2-Ethylthio-1,3,4-thiadiazole-5-thiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.