missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylthio)ethanol, TRC

Product Code. 30372272
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372272 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372272 Supplier Toronto Research Chemicals Supplier No. TRCE9267801G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 110-77-0
Molecular Formula C4H10OS
Quantity 1 g
Synonym 2-Ethylmercaptoethanol, 2-Hydroxyethyl Ethyl Sulfide, Ethyl 2-Hydroxyethyl Sulfide, Ethyl 2-Hydroxyethyl Thioether, Ethyl β-Hydroxyethyl Sulfide, NSC 57105, β-Ethylthioethanol, β-Hydroxydiethyl Sulfide
InChI Formula InChI=1S/C4H10OS/c1-2-6-4-3-5/h5H,2-4H2,1H3
SMILES OCCSCC
Molecular Weight (g/mol) 106.19
Chemical Name or Material 2-(Ethylthio)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.