missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyltoluene, 98+%

Product Code. 11361487
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11361487 Supplier Thermo Scientific Alfa Aesar Supplier No. L02028.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-14-3 | C9H12 | 120.195 g/mol

2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.

Solubility
Soluble in water (74.6 mg/L (25°C)).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-14-3
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00009257
InChI Key HYFLWBNQFMXCPA-UHFFFAOYSA-N
Synonym 2-ethyltoluene, o-ethyltoluene, o-methylethylbenzene, 1-methyl-2-ethylbenzene, toluene, o-ethyl, ortho-ethyltoluene, benzene, 1-ethyl-2-methyl, 1,2-methylethylbenzene, o-ethyl methylbenzene, o-ethyl methyl benzene
PubChem CID 11903
ChEBI CHEBI:34276
IUPAC Name 1-ethyl-2-methylbenzene
SMILES CCC1=CC=CC=C1C

Specifications

Melting Point -17°C
Density 0.884
Boiling Point 164°C to 165°C
Flash Point 53°C (127°F)
Refractive Index 1.504
Quantity 5 g
UN Number UN3295
Beilstein 1851237
Solubility Information Soluble in water (74.6mg/L (25°C)).
Formula Weight 120.2
Percent Purity ≥98%
Chemical Name or Material 2-Ethyltoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.