missing translation for 'onlineSavingsMsg'
Learn More

2-Ethynylpyridine, 98+%

Product Code. 15119263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15119263 25 g Glass bottle 25g
10297030 1 g Glass bottle 1g
10750595 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15119263 Supplier Thermo Scientific Acros Supplier No. 295670250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1945-84-2 | C7H5N | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1945-84-2
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.12
MDL Number MFCD00041598
InChI Key NHUBNHMFXQNNMV-UHFFFAOYSA-N
Synonym 2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine
PubChem CID 137268
IUPAC Name 2-ethynylpyridine
SMILES C#CC1=CC=CC=N1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Brown
Density 1.0200g/mL
Boiling Point 77.0°C to 78.0°C (14.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5570 to 1.5610
Quantity 25 g
Specific Gravity 1.02
Solubility Information Solubility in water: soluble in water
Formula Weight 103.12
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Ethynylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.