missing translation for 'onlineSavingsMsg'
Learn More

2-Ethynylthiophene, ≥90%, Thermo Scientific™

Product Code. 10792984
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10792984 250 mg Amber Glass Bottle 250mg
10104053 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10792984

Brand: Thermo Scientific Maybridge CC13047CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4298-52-6
Molecular Formula C6H4S
Molecular Weight (g/mol) 108.16
MDL Number MFCD03425129
InChI Key LWISLHRIEATKTM-UHFFFAOYSA-N
Synonym 2-ethynyl-thiophene, 2-thienylethine, 2-thiophenylacetylene, 2-thienyl acetylene, pubchem16123, thien-2-yl acetylene, thiophene, 2-ethynyl, thiophen-2-yl acetylene, 2-ethynylthiophene, acmc-209js9
PubChem CID 5246258
IUPAC Name 2-ethynylthiophene
SMILES C#CC1=CC=CS1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 108.16
Percent Purity 90%
Chemical Name or Material 2-ethynylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.