missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-1,3-dimethylbenzene, 97%, Thermo Scientific™

Product Code. 10458002
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10458002 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10458002 Supplier Thermo Scientific Maybridge Supplier No. CD05584DA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 443-88-9
CAS Min % 96.5
CAS Max % 100.0
Density 0.988g/mL
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H9F
Linear Formula (CH3)2C6H3F
MDL Number MFCD00039215
Synonym 2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene
InChI Key JTAUTNBVFDTYTI-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)C)F
IUPAC Name 2-fluoro-1,3-dimethylbenzene
Molecular Weight (g/mol) 124.158
PubChem CID 123065
Formula Weight 124.16
Percent Purity 97%
Chemical Name or Material 2-Fluoro-1, 3-dimethylbenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.