missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals

Product Code. 15486899
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15486899 Supplier Thermo Scientific Alfa Aesar Supplier No. J65441.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64183-27-3 | Molecular Formula: C10H12FN5O3 | Molecular Weight: 269.24 g/mol

2'-Fluoro-2'-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2'-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-2′-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2′-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64183-27-3
Molecular Formula C10H12FN5O3
Molecular Weight (g/mol) 269.24
MDL Number MFCD09750859
InChI Key ZGYYPTJWJBEXBC-GPGUAWMRNA-N
Synonym 2'-fluoro-2'-deoxyadenosine, 2'-deoxy-2'-fluoroadenosine, adenosine, 2'-deoxy-2'-fluoro, 2r,3r,4r,5r-5-6-amino-9h-purin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 9-2-fluoro-2-deoxy-beta-d-ribofuranosyl-purin-6-amine, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl oxolan-3-ol, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 2-fluoro-2-deoxyadenosine, 2'f-da
PubChem CID 100253
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3F)C2=NC=N1

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Formula Weight 269.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.