missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Fluoro-2'-deoxyguanosine, 99%

Product Code. 15436179
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436179 1 g 1g
15446179 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436179 Supplier Thermo Scientific Chemicals Supplier No. J64055.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 78842-13-4 | C10H12FN5O4 | 285.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 78842-13-4
Molecular Formula C10H12FN5O4
Molecular Weight (g/mol) 285.24
MDL Number MFCD00923832
InChI Key UXUZARPLRQRNNX-SJLGBTOWNA-N
Synonym 2'-deoxy-2'-fluoroguanosine, 2'-fluoro-2'-deoxyguanosine, guanosine, 2'-deoxy-2'-fluoro, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl-tetrahydro-furan-2-yl-1,9-dihydro-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 2'-dfgs, deoxyfluoroguanosine, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one
PubChem CID 196536
IUPAC Name 2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F)C(=O)N1

Specifica

Color White
Quantity 1 g
Formula Weight 285.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxyguanosine

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.