missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-2'-deoxyuridine, 97%

Product Code. 15284418 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15284418 5 g 5g
15274418 1 g 1g
15294418 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15284418 Supplier Thermo Scientific Acros Supplier No. 455000050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 784-71-4 | C9H11FN2O5 | 246.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 784-71-4
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD01317293
InChI Key UIYWFOZZIZEEKJ-XVFCMESISA-N
Synonym 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203
PubChem CID 150851
IUPAC Name 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Melting Point 151.0°C
Loss on Drying 1% max. (1 g, 105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 246.19
Percent Purity 97%
Chemical Name or Material 2'-Fluoro-2'-deoxyuridine
compliance-icons compliance-icons
Product Identifier
  • 2'-Fluoro-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.