missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Fluoro-2'-deoxyuridine, 97%

Product Code. 15497139
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497139 25 g 25g
15477139 1 g 1g
15487139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497139 Supplier Thermo Scientific Chemicals Supplier No. J65948.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 784-71-4 | C9H11FN2O5 | 246.19 g/mol

2'-Fluoro-2'-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-2′-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.

Solubility
Soluble in methanol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 784-71-4
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD01317293
InChI Key UIYWFOZZIZEEKJ-XVFCMESISA-N
Synonym 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203
PubChem CID 150851
IUPAC Name 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Color White
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 246.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxyuridine
compliance-icons compliance-icons
Product Identifier
  • 2'-Fluoro-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2'-Fluoro-2'-deoxyuridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.