missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific™

Product Code. 15127422 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Product Code. Quantity Packaging unitSize
15127422 5 g Glass bottle 5g
15147422 25 g Glass bottle 25g
15157422 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 15127422

Brand: Thermo Scientific Acros 263540050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 74457-86-6
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD00042290
InChI Key PIRRWUMTIBFCCW-UHFFFAOYSA-N
Synonym 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344
PubChem CID 592821
IUPAC Name 1-(2-fluoro-4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C(C)=O)C(F)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 54.0°C
Color Colorless
Boiling Point 80.0°C to 85.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 168.17
Percent Purity 99%
Physical Form Viscous Solution
Chemical Name or Material 2'-Fluoro-4'-methoxyacetophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Fluoro-4'-methoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.