missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-hydroxypyridine, 95%, Thermo Scientific Chemicals

Product Code. 11312889
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
11312889 1 g 1g
11322889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11312889 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H27741.03
 Restricted Item
Estimated Shipment: 19-10-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format
Denna artikel kan inte returneras. Se returpolicy

CAS: 55758-32-2 | Molecular Formula: C5H4FNO | Molecular Weight: 113.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 55758-32-2
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.091
MDL Number MFCD01075040
InChI Key HTRLNWYWOKWCLV-UHFFFAOYSA-N
Synonym 2-fluoro-5-hydroxypyridine, 6-fluoro-3-pyridinol, 6-fluoro-3-hydroxypyridine, 3-pyridinol, 6-fluoro, pubchem6321, 6-fluoropyridine-3-ol, 6-fluoro-pyridin-3-ol, acmc-1b21k, 3-hydroxy-6-fluoro-pyridine, ksc490s4n
PubChem CID 2763108
IUPAC Name 6-fluoropyridin-3-ol
SMILES C1=CC(=NC=C1O)F

Specifikationer

Melting Point 150°C to 153°C
Quantity 1 g
Formula Weight 113.09
Percent Purity 95%
Chemical Name or Material 2-Fluoro-5-hydroxypyridine

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.