missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitroaniline, 98%

Product Code. 10321034 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10321034 5 g Glass bottle 5g
10285880 25 g Glass bottle 25g
10377870 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10321034 Supplier Thermo Scientific Acros Supplier No. 168390050
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 369-36-8 | C6H5FN2O2 | 156.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-36-8
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007652
InChI Key KJVBJICWGQIMOZ-UHFFFAOYSA-N
Synonym benzenamine, 2-fluoro-5-nitro, 2-fluoro-5-nitrophenylamine, 2-fluoro-5-nitro-phenylamine, 1-amino-2-fluoro-5-nitrobenzene, 2-fluoro-5-nitrobenzenamine, 2-fluoro-5-nitro aniline, 3-amino-4-fluoronitrobenzene, pubchem8504, acmc-1crtl, 5-nitro-2-fluoroaniline
PubChem CID 67785
IUPAC Name 2-fluoro-5-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 99°C to 103°C
Color Brown to Ochre-Yellow
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H3(NO2)NH2
Quantity 5 g
Beilstein 12, I, 355
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,alcohol and toluene
Formula Weight 156.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-nitroaniline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-5-nitroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.