missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methylnaphthalene, 97%, Maybridge

Product Code. 10642843
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10642843 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10642843 Supplier Thermo Scientific Maybridge Supplier No. RH01072CB
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 324-42-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H9F
MDL Number MFCD00831714
Quantity 250 mg
Synonym 6-fluoro-2-methylnaphthalene, naphthalene, 2-fluoro-6-methyl, maybridge1_006492, 2-methyl-6-fluoronaphthalene, naphthalene,2-fluoro-6-methyl
InChI Key IKKGPHTUCGWMPE-UHFFFAOYSA-N
SMILES CC1=CC=C2C=C(F)C=CC2=C1
IUPAC Name 2-fluoro-6-methylnaphthalene
Molecular Weight (g/mol) 160.19
PubChem CID 2727845
Formula Weight 160.19
Percent Purity 97%
Chemical Name or Material 2-Fluoro-6-methylnaphthalene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.