missing translation for 'onlineSavingsMsg'
Learn More

2-fluoro-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline-11-carboxylic acid, Thermo Scientific™

Product Code. 10233092
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10233092 1 g 1g
10089501 250 mg 250mg
10230415 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10233092 Supplier Thermo Scientific Maybridge Supplier No. SEW05649DA
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1555-11-9
Molecular Formula C15H14FNO2
Molecular Weight (g/mol) 259.28
InChI Key HBSUFSHHGHJZSE-UHFFFAOYSA-N
Synonym 2-fluoro-7,8,9,10-tetrahydro-6h-cyclohepta b quinoline-11-carboxylic acid, 2-fluoro-6h,7h,8h,9h,10h-cyclohepta b quinoline-11-carboxylic acid, maybridge1_008193, 2-fluoranyl-7,8,9,10-tetrahydro-6h-cyclohepta b quinoline-11-carboxylic acid, 2-fluoro-6,7,8,9,10-pentahydrocyclohepta 1,2-b quinoline-11-carboxylic acid, 6h-cyclohepta b quinoline-11-carboxylic acid, 2-fluoro-7,8,9,10-tetrahydro
PubChem CID 715240
IUPAC Name 2-fluoro-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline-11-carboxylic acid
SMILES C1CCC2=C(CC1)N=C3C=CC(=CC3=C2C(=O)O)F

Specifications

Quantity 1 g
Formula Weight 259.28
Percent Purity 97%
Chemical Name or Material 2-fluoro-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline-11-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.