missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-9-beta-D-arabinofuranosyladenine, 98%

Product Code. 15426969
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426969 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426969 Supplier Thermo Scientific Alfa Aesar Supplier No. J65562.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21679-14-1 | C10H12FN5O4 | 285.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21679-14-1
Color White to Yellow
Molecular Formula C10H12FN5O4
MDL Number MFCD00132942
Quantity 1 g
Beilstein 1225932
Merck Index 14,4126
Synonym fludarabine, f-ara-a, 2-fluoro ara-a, 9-beta-d-arabinofuranosyl-2-fluoroadenine, fludarabine inn, fludarabinum latin, fludarabina spanish, fludarabina, fludarabinum, 2r,3s,4s,5r-2-6-amino-2-fluoro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol
Solubility Information Soluble in DMF, DMSO, methanol or ethanol; Sparingly soluble in water
InChI Key HBUBKKRHXORPQB-FJFJXFQQSA-N
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O)C2=NC(F)=N1
IUPAC Name (2R,3S,4S,5R)-2-(6-amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 285.24
PubChem CID 657237
Formula Weight 285.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-9-beta-D-arabinofuranosyladenine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.