Learn More
Thermo Scientific™ TheraPure™ 2'-Fluoro-Ac-C Phosphoramidite, 20-400 finish bottle
Shop All Thermo Scientific ProductsDescription
2'-Fluoro-Ac-C CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with 2'-Fluoro protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, the highly electronegative fluorine (2'-Fluoro) modification adopts a more thermodynamically stable A-form helix on hybridization to its target. Showing improved affinity and some nuclease resistance (compared to 2'-OH), the 2'-F modification has been proven useful in antisense, siRNA, and aptamer applications. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.
Specifications
Specifications
| 31P NMR Purity | ≥98% |
| Format | Amber bottle (20-400 finish neck style) |
| For Use With (Equipment) | Expedite Synthesizers |
| Description | Base: C Base Protection: Ac |
| Molecular Weight (g/mol) | 789.8 g/mol |
| Product Type | 2in-Fluoro-Ac-C CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Cytidine (C) |
| Reagent Features | 2in-Fluoro modified, Fast deprotection |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.