missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-alpha-methyl-4-biphenylacetic acid, 99%

Produktkod. 11455187
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
11455187 5 g 5g
11445187 1 g 1g
11465187 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11455187 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' B22603.06
 Restricted Item
Estimated Shipment: 10-09-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format
Denna artikel kan inte returneras. Se returpolicy

CAS: 5104-49-4 | C15H13FO2 | 244.27 g/mol

A cyclooxygenase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 5104-49-4
Molecular Formula C15H13FO2
Molecular Weight (g/mol) 244.27
MDL Number MFCD00079303
InChI Key SYTBZMRGLBWNTM-UHFFFAOYNA-N
Synonym flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
PubChem CID 3394
ChEBI CHEBI:5130
SMILES CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

Specifikationer

Melting Point 112°C to 114°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Merck Index 14,4199
IUPAC Name 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Formula Weight 244.27
Percent Purity 99%
Chemical Name or Material 2-Fluoro-alpha-methyl-4-biphenylacetic acid

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.