Learn More
Thermo Scientific™ TheraPure™ 2'-Fluoro-U Phosphoramidite, 20-400 finish bottle
Shop All Thermo Scientific ProductsBeschreibung
2'-Fluoro-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-Fluoro protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, the highly electronegative fluorine (2'-Fluoro) modification adopts a more thermodynamically stable A-form helix on hybridization to its target. Showing improved affinity and some nuclease resistance (compared to 2'-OH), the 2'-F modification has been proven useful in antisense, siRNA, and aptamer applications. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-fluorouridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C39H46FN4O8P
• Base: A
• Formula weight: 748.8 g/mol
• CAS number: 146954-75-8
• HPLC purity: >98%
• 31P NMR purity: ≥98%
Spezifikation
Spezifikation
| For Use With (Equipment) | Expedite Synthesizers |
| Product Type | 2in-Fluoro-U CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Uridine (U) |
| Reagent Features | 2in-Fluoro modified |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 0.25 g |
| Reagent Type | Phosphoramidite |
| Grade | TheraPure™ |
| Mehr anzeigen |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.