missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoroacetophenone, 97%

Product Code. 11452098
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452098 100 g 100g
11432098 5 g 5g
11442098 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452098 Supplier Thermo Scientific Alfa Aesar Supplier No. A15128.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 445-27-2 | C8H7FO | 138.141 g/mol

2'-Fluoroacetophenone is used as starting reagent in the synthesis of ascididemin. It is used to produce 1-(2-piperidin-1-yl-phenyl)-ethanone by reaction with piperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoroacetophenone is used as starting reagent in the synthesis of ascididemin. It is used to produce 1-(2-piperidin-1-yl-phenyl)-ethanone by reaction with piperidine.

Solubility
Soluble in Acetone, Chloroform, Dichloromethane, Ethanol, Ethyl Acetate and Methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-27-2
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.141
MDL Number MFCD00000320
InChI Key QMATYTFXDIWACW-UHFFFAOYSA-N
Synonym 2'-fluoroacetophenone, 1-2-fluorophenyl ethanone, o-fluoroacetophenone, 1-2-fluorophenyl ethan-1-one, 1-acetyl-2-fluorobenzene, ethanone, 1-fluorophenyl, 2-fluoroacetophenone, 2'fluoroacetophenone, 2;-fluoroacetophenone
PubChem CID 96744
IUPAC Name 1-(2-fluorophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1F

Specifications

Density 1.137
Boiling Point 82°C to 83°C (10 mmHg)
Flash Point 61°C (141°F)
Refractive Index 1.5075
Quantity 100 g
Beilstein 2041344
Solubility Information Soluble in Acetone,Chloroform,Dichloromethane,Ethanol,Ethyl Acetate and Methanol.
Formula Weight 138.14
Percent Purity 97%
Chemical Name or Material 2'-Fluoroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.