missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzal Chloride, TRC

Product Code. 30390445
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390445 1 g
30389119 100 mg
30395356 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390445 Supplier Toronto Research Chemicals Supplier No. TRCF1256001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-65-0
Molecular Formula C7H5Cl2F
Molecular Weight (g/mol) 179.02
Synonym o-Fluorobenzal Chloride, 2-Fluoro(dichloromethyl)benzene, NSC 60714, 1-Dichloromethyl-2-fluorobenzene, 1-(Dichloromethyl)-2-fluorobenzene
IUPAC Name 1-(dichloromethyl)-2-fluorobenzene
SMILES Fc1ccccc1C(Cl)Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5Cl2F/c8-7(9)5-3-1-2-4-6(5)10/h1-4,7H
Formula Weight 177.9752
Chemical Name or Material 2-Fluorobenzal Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.