missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzeneacetic Acid, TRC

Product Code. 30397803
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397803 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397803 Supplier Toronto Research Chemicals Supplier No. TRCF588310100G
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Fluorobenzeneacetic Acid
CAS 451-82-1
Molecular Formula C8H7FO2
Quantity 100 g
Synonym (o-Fluorophenyl)acetic Acid, (2-Fluorophenyl)acetic Acid, (o-Fluorophenyl)acetic Acid, 2-(2-Fluorophenyl)ethanoic Acid, NSC 401, o-Fluorobenzeneacetic Acid
InChI Formula InChI=1S/C8H7FO2/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4H,5H2,(H,10,11)
SMILES FC1=CC=CC=C1CC(O)=O
IUPAC Name 2-(2-fluorophenyl)acetic acid
Molecular Weight (g/mol) 154.14
Formula Weight 154.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.