missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzenesulfonyl chloride, 97%

Product Code. 10105832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10105832 1 g Glass bottle 1g
10619784 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10105832

Brand: Thermo Scientific Acros 375780010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-21-7 | C6H4ClFO2S | 194.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-21-7
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00042285
InChI Key ZSZKAQCISWFDCQ-UHFFFAOYSA-N
Synonym 2-fluorobenzene-1-sulfonyl chloride, 2-fluorobenzenesulphonyl chloride, 2-fluorobenzenesulfonylchloride, 2-fluoro-benzenesulfonyl chloride, 2-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 2-fluoro, chloro 2-fluorophenyl sulfone, pubchem5138, ksc205g6f
PubChem CID 137761
IUPAC Name 2-fluorobenzenesulfonyl chloride
SMILES FC1=CC=CC=C1S(Cl)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 26.0°C
Color Colorless
Boiling Point 140.0°C to 142.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula FC6H4SO2Cl
Refractive Index 1.5350 to 1.5380
Quantity 1 g
Formula Weight 194.61
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.