missing translation for 'onlineSavingsMsg'
Learn More

2-(Fmoc-amino)isobutyric acid, 98%, Thermo Scientific Chemicals

Product Code. 15458758
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458758 25 g 25g
15438758 1 g 1g
15448758 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458758 Supplier Thermo Scientific Alfa Aesar Supplier No. H62211.14
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94744-50-0
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.36
MDL Number MFCD00151913
InChI Key HOZZVEPRYYCBTO-UHFFFAOYSA-N
Synonym fmoc-aib-oh, fmoc-alpha-methylalanine, fmoc-alpha-me-ala-oh, 2-fmoc-amino isobutyric acid, 2-9h-fluoren-9-yl methoxy carbonyl amino-2-methylpropanoic acid, alanine, n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl, 2-9h-fluoren-9-ylmethoxy carbonyl amino-2-methylpropanoic acid, n-fmoc-2-aminoisobutyric acid, 2-9h-fluoren-9-ylmethoxy carbonylamino isobutyric acid
PubChem CID 2756096
SMILES CC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 182°C to 188°C
Quantity 25 g
Beilstein 5604328
Sensitivity Moisture sensitive
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-methylpropanoic acid
Formula Weight 325.36
Percent Purity 98%
Chemical Name or Material 2-(Fmoc-amino)isobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.